Laboratory Micro Burner
Supplier: GLOBAL MARKET CONNECTIONS, LTD
EW-M1 EW-M1 MF-2001
470150-450CS 1304.9 CAD
470150-450 470199-856 470202-758
Laboratory Micro Burner
Heaters Gas Burners
Safe and portable option to using Bunsen burner.
  • No wicks or gas lines to install
  • Fills quickly and easily from a butane cartridge, which is available separately
It features a stable, tip-resistant design, push button auto ignition, and an adjustable flame size. Size: 3"H x 3" dia.
Accessories information: A safe and easy way to refill the micro burner (15 V 0906), this refill stand leaves both hands free to insert the butane canister into the valve on bottom of burner. Simply invert your micro burner in the stand and fill it easily without getting your hands cold or spilling fuel.
Other available accessories for Laboratory Microburner:
• Tripod Stand, 15 V 0731
• Accessory Kit (wire gauze pad, evaporating dish holder, three clay triangles), 15 V 0915
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.