Laboratory Jacks
Supplier: United Scientific Supplies
LBJK01 LBJSET LBJSET
470001-938EA 237.7 CAD
470001-938 470006-698 470199-870
Laboratory Jacks
Laboratory Jacks
Laboratory support jack is constructed of painted aluminum and stainless steel.
  • Scissor jack design enables height adjustment with a simple turn of the knob
  • Sturdy design can support loads up to 20 kg
  • Platform height can be vertically adjusted from closed position of 2.75" up to 10.5"
  • Pre-drilled holes for attaching a support rod
Available as a set with removable support rod included.