Economy Digital and Analog Incubators
Supplier: Quincy Labs
10-140 12-140E 12-140 10-140E
470006-400EA 1170.3 CAD
470006-400 470104-838 470001-908 470020-304
Economy Digital and Analog Incubators
Incubators Compact Benchtop Incubators
Economy gravity convection ovens feature a perforated heat shield for good temperature uniformity for quick run-up and recovery times.
  • Incubator dries, bakes, sterilizes, and evaporates
  • Maximum temperature: 62°C (145°F)
  • Scratch-resistant finish
  • Dual-range laboratory thermometer
  • Three-wire cord and plug
  • Aligned with GLOBE protocols
They also have heavy-steel, double-wall construction and a corrosive-resistant, aluminized steel interior.
The light gray hammer finish is scratch resistant. It has two adjustable chrome-plated wire shelves and supports.
More Product Information