Kerosene

Supplier: ALDON CORP SE

KK0035-1L KK0035-500ML
470301-488EA 72.45 CAD
470301-488 470301-490
Kerosene
Chemicals for Science Education

CAS Number: 64741-65-7
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Flammable
Density (g/mL): 0.758
Boiling Point (°C): 175-195
Solubility: Many Organic Solvents
Synonyms: Naphtha (Petroleum), Heavy Alkylate
Shelf Life (months): 36
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR