Isopropyl Alcohol

Supplier: ALDON CORP SE

IX0230-500ML IX0230-PK/2 IX0235-1LR IX0235-3.8LR IX0235-500MLR IX0235-1L IX0235-3.8L IX0235-500ML IX0235-3.8SCR IX0235-3.8SC IX0235-250ML IX0235-20L IX0235-2/PK IX0230-3.8L IX0229-250ML
470301-456EA 16.85 CAD
470301-456 470301-458 470301-460 470301-464 470301-466 470301-468 470301-474 470301-476 470301-462 470301-472 470301-480 470301-756 470301-772 470157-450 470225-824
Isopropyl Alcohol
Chemicals for Science Education

CAS Number: 67-63-0
Formula: CH32CHOH
Density: 0.786—0.79 g/mL
Boiling and Freezing Point: 82°C, -90°C
Solubility: Water, Alcohol and Ether
Synonyms: 2-Propanol, Isopropanol
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable and toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR