Isopentyl Alcohol

Supplier: ALDON CORP SE

IX0225-100MLR IX0225-500MLR
470301-722EA 37.05 CAD
470301-722 470301-724
Isopentyl Alcohol
Chemicals for Science Education

CAS Number: 123-51-3
Formula Weight: 88.2
Formula: (CH3)2CHCH2CH2OH
Hazard Info: Flammable, Toxic, Irritant
Density (g/mL): 0.813-0.816
Boiling Point (°C): 133
Freezing Point (°C): -118
Solubility: Slightly in Water
Synonyms: Isoamyl Alcohol, 3-Methyl-1-Butanol
Shelf Life (months): 12
Storage: Red


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR