Iron(III) Oxide Red

Supplier: ALDON CORP SE

FF0120-500GR FF0120-100G FF0120-500G
470301-418EA 59.55 CAD
470301-418 470301-414 470301-648
Iron(III) Oxide Red
Chemicals for Science Education

CAS Number: 1309-37-1
Formula Weight: 159.68
Formula: Fe2O3
Density (g/mL): 5.24
Freezing Point (°C): 1565
Solubility: Acids
Synonyms: Ferric Oxide Red
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR