Iron(III) Nitrate Nonahydrate

Supplier: ALDON CORP SE

FF0110-100GR FF0110-500GR FF0112-50ML FF0110-100G FF0110-500G FF0110-PK/4R FF0112-500ML FF0113-CONC FF0121-500ML FF0122-500ML
470301-404EA 22.9 CAD
470301-404 470301-408 470301-398 470301-400 470301-402 470301-406 470301-410 470301-412 470225-700 470225-702
Iron(III) Nitrate Nonahydrate
Chemicals for Science Education

CAS Number: 7782-61-8
Formula: FeNO33 9H2O
Density: 1.68 g/mL
Freezing Point: 47.2°C
Solubility: Water and Alcohol
Synonyms: Iron (III) Nitrate 9-Hydrate, Ferric Nitrate Nonahydrate
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is an irritant and oxidizer.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR