Iron(III) Chloride

Supplier: ALDON CORP SE

FF0100-100G FF0100-100GR FF0100-500GR FF0101-500ML FF0102-100ML FF0102-CONC FF0100-500G
470301-382EA 30 CAD
470301-382 470301-386 470301-388 470301-392 470301-394 470301-396 470301-584
Iron(III) Chloride
Chemicals for Science Education

CAS Number: 7705-08-0
Formula Weight: 162.21
Formula: FeCl3
Hazard Info: Irritant, Corrosive, Hygroscopic
Density (g/mL): 2.80-2.90
Boiling Point (°C): 307
Freezing Point (°C): 307
Solubility: Water
Synonyms: Ferric Chloride Anhydrous
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR