Iron(III) Chloride Hexahydrate

Supplier: ALDON CORP SE

FF0080-100GR FF0080-500GR FF0080-PK/4R FF0080-100G FF0080-500G
470301-378EA 35.95 CAD
470301-378 470301-380 470301-390 470301-564 470301-568
Iron(III) Chloride Hexahydrate
Chemicals for Science Education

CAS Number: 10025-77-1
Formula Weight: 270.30
Formula: FeCl3·6H2O
Hazard Info: Irritant, Corrosive
Freezing Point (°C): 37
Solubility: Water and Common Organic Solvents
Synonyms: Ferric Chloride Hexahydrate, Iron(III) Chloride 6-Hydrate
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR