Iron(II) Sulfate Heptahydrate

Supplier: ALDON CORP SE

FF0190-500G FF0190-100GR FF0190-500GR FF0190-2.5KG FF0152-500ML
470301-362EA 27 CAD
470301-362 470301-364 470301-366 470301-536 470225-752
Iron(II) Sulfate Heptahydrate
Chemicals for Science Education

CAS Number: 7782-63-0
Formula Weight: 278.02
Formula: FeSO4·7H2O
Hazard Info: Toxic
Density (g/mL): 1.897
Boiling Point (°C): Decomposes
Freezing Point (°C): 72
Solubility: Water
Synonyms: Ferrous Sulfate 7-Hydrate, Iron(II) Sulfate 7-Hydrate, Ferrous Sulfate Heptahydrate
Shelf Life (months): 12
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR