Ward's® Introductory Mineral and Rock Collection
Supplier: Avantor
Ward's®
WARD470025-166
470025-166EA 69.29 CAD
470025-166
Ward's® Introductory Mineral and Rock Collection
Specimens Rock Specimens Rock Collections
Examples of the most basic rocks and rock-forming minerals.
  • Sixteen 1×2" numbered samples
  • Housed in a compartmented collection box
  • Identification key included
Give students a clear introduction to rocks and rock-forming minerals; the specimens in the introductory set are easy to differentiate from one another and clearly show the traits of each type.
Minerals: Biotite, feldspar, quartz, calcite
Igneous: Basalt, granite, rhyolite, gabbro
Sedimentary: Conglomerate, limestone, sandstone, shale
Metamorphic: Gneiss, marble, quartzite, schist