Introduction to Qualitative Analysis of Proteins and Amino Acids Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
8-150
470008-066KT 467.1 CAD
470008-066
Introduction to Qualitative Analysis of Proteins and Amino Acids Lab Activity
Educational Materials Biology Educational Materials Biochemistry Learning Activities
This kit demonstrates the principles of protein and amino acid analysis and enables students to identify unknown compounds.
  • Activities Connect Chemistry and Biology
  • Students Test for Proteins and Amino Acids
  • Amino Acids Included: Glutamic Acid, Glycine, Cystine, and More
  • Proteins Included: Albumin and Gelatin
  • Purpose for Students: to Learn How to Identify Unknown Compounds
In this set of lab activities, students work with the proteins albumin and gelatin and the amino acids glutamic acid, glycine, cystine, cysteine, tyrosine, tryptophan, and arginine. Ninhydrin detects the presence of amines and indicates the presence of amino acids and/or proteins, the Biuret test detects the presence of peptide bonds, and specific analytical tests determine the presence of individual amino acids.
Protein and amino acid analysis techniques are an integral part of biochemistry, and the range of tests available for analyzing amino acids and proteins indicate their importance.
Ordering information: This kit includes a pipet, biuret solution, sodium hypochlorite solution, alpha-naphthol solution, sodium hydroxide (three different solutions), ninhydrin, lead acetate, albumin, gelatin, glutamic acid, glycine, cysteine, arginine, DL tyrosine, DL tryptophan, cystine, unknown amino acid, protein-testing solution B, and sodium hydroxide. Concentrated nitric acid, ethanol, hot plates, balances, glassware, and distilled water are required but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.