Explore the pivotal role that enzymes play in the metabolic process.
- Labs show the activity of invertase, amylase, and more
- Material for six student groups
Study the enzyme activity of invertase, amylase, and pepsin using specific enzyme assay systems. Kit includes five introductory experiments: demonstrating invertase activity, amylase activity, pepsin activity, the presence of amylase in saliva, and the effect of time on the activity of invertase. Invertase and amylase activity are measured using a dinitrosalicylic acid color change system. Enzyme reaction rates may be measured by visual examination or spectrophotometry. Includes all required chemicals and a prepaid reply card for Invertase, amylase, pepsin and hemoglobin to be shipped separately upon request within the continental United States. Activities are appropriate for advanced biology or biochemistry courses.
Ordering information: This set includes data sheets, material for six student groups, and a teacher's manual with suggestions for lab situations with special needs students. It also includes assessment manuals and scoring sheets.
This item contains a coupon for perishable materials that must be redeemed from the manufacturer before use. Please allow sufficient time to account for redemption. These materials are shipped from the supplier via second day delivery and must be refrigerated upon receipt.
Delivery information: See datasheet for contents information.