Ward's® Introduction to Covalent Molecules Lab Activity
Supplier: Avantor
WARD470007-576
470007-576KT 167.29 CAD
470007-576 470328-439
Ward's® Introduction to Covalent Molecules Lab Activity
Models Molecular Biology Models
Basic Molecular Structure Shown in Three-Dimensional Model.
  • Visualize the Electron-Dot diagrams
  • Teacher's guide included
  • Aligned to NGSS standards (MS/HS: PS1, PS2, PS3)
Teach students how to transfer the concept of basic molecular structure from a two-dimensional electron-dot diagram to an actual three-dimensional model in a few easy steps. The three-dimensional models provide a realistic representation of the configuration of a substance and indicate its physical properties. Studying the electron-dot diagram and then building a molecular model allows students to more easily grasp how unshared electron pairs and covalent bonds determine the structure of molecules.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.