Intestinal Juice, Simulated

Supplier: ALDON CORP SE
IX0110-100ML
470301-304EA 26.55 CAD
470301-304
Intestinal Juice, Simulated
Chemicals for Science Education
Hazard Info: Irritant
Solubility: Water
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR