InstaStain® Methylene Blue
Supplier: Edvotek
2003
2004
2004
470002-528EA
162.6
CAD
470002-528
470002-530
470200-110
InstaStain® Methylene Blue
Chemicals for Science Education
New and improved, one-step staining and destaining.
Stain DNA gels quickly and efficiently with environmentally safe InstaStain®. Minimize mess and liquid waste, while yielding high quality uniform gel results excellent for photodocumentation. Each 7x7cm card stains one gel.