Ward's® Instant Drosophila Medium
Supplier: Avantor
470015-394PK 79.95 CAD
470015-394 470015-396 470024-736 470024-738 470024-740 470024-742
Ward's® Instant Drosophila Medium
Specimens Live Specimens Animals - Living Material
Improved Formula for Higher Yields
This ready-to-use formula, developed by WARD’S, just needs water. No cooking, sterilization, or additional inhibitors are necessary to create the perfect environment for culturing Drosophila. In addition, the medium won’t soften or liquefy, so flies won’t drown. Choose white or blue for better observation of developing larvae. The color won’t harm your Drosophila. One liter is sufficient for approximately 60 standard cultures.