Innoculating Loops
Supplier: United Scientific Supplies
INL703 3412 INL703-PK/4
470123-080EA 9.65 CAD
470123-080 470203-988 470321-002
Innoculating Loops
Spreaders Inoculating Loops and Needles
Color set of four inoculating loops with brass handles and four colors.
  • Non-insulated aluminum handle
Twisted 3" lengths of non-calibrated 26 gauge nickel-chromium wire with 8 mm loops set in threaded aluminum handles. Wire is non-replaceable.