Boreal Science Compound Beginner Microscopes
Supplier: VWR
Boreal
5791200 5585000
470177-096EA 329.2 CAD
470177-096 470177-094
Boreal Science Compound Beginner Microscopes
Microscopes
Boreal beginner microscopes are ideal for introducing students to microscopy.
  • All-Glass-Coated Optics
  • All-Metal Construction
  • Parfocal and Parcentered
  • Objectives: 4x, 10x, and 40x
  • Boreal Lifetime Warranty
It's scaled down for younger students and is simple to use, but it also prepares students for the transition to high school instruction.
Rugged metal components are engineered for years of reliable performance. Student-proof features minimize loss of parts.
Ordering information: This product includes a manufacturer's warranty.