Ward's® Igneous Rock Collection
Supplier: Avantor
470154-094EA 277.25 CAD
470154-094 470025-222
Ward's® Igneous Rock Collection
Specimens Rock Specimens Igneous Rocks
Samples of Intrusive and Extrusive Igneous Rocks
  • Introduce concepts of the Earth's crust
  • Compartmented box
Provide your students with an excellent introduction to the forces at work beneath the Earth’s crust. The set is housed in a compartmented collection box and includes an identification list.
Contents:
12 numbered samples, 1" x 2"
Rocks:
basalt, diorite, gabbro, granite, obsidian, pegmatite, peridotite, pumice, rhyolite (2), scoria, syenite
Hand Specimen Set:
Large hand samples are ideal for demonstration and display. This set is housed inMore Product Information