Hydrochloric Acid

Supplier: ALDON CORP SE

HH0100-1L HH0093-500ML HH0090-1L HH0090-3.8L HH0090-500ML HH0090-PK2 HH0095-500ML HH0170-500ML HH0170-PK/2 HH0158-3ML HH0171-2.5LR HH0171-100MLR HH0171-500MLR HH0071-CONC HH0100-3.8L HH0093-1L HH0120-1L HH0094-500ML HH0168-1L HH0168-3.8L HH0170-1L HH0171-PK/2R HH0171-PK/4R HH0171-PK2R HH0071-2.5L HH0099-1L AD-HH0171P-2.5L AD-HH0171P-PK/6
470113-126EA 14.69 CAD
470113-126 470301-224 470301-228 470301-232 470301-234 470301-236 470301-238 470301-240 470301-250 470301-252 470301-254 470301-256 470301-260 470301-264 470301-218 470301-222 470301-226 470301-230 470301-242 470301-244 470301-246 470301-248 470301-258 470301-262 470301-266 470301-268 470225-696 470354-386 470354-380
Hydrochloric Acid
Chemicals for Science Education

CAS Number: 7647-01-0
Formula: HCl
Density: 1.16 mL
Boiling and Freezing Point: 81.11 to 85°C, -45°C
Solubility: Water
Synonyms: Muriatic Acid, Hydrogen Chloride
Shelf Life: 24 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is corrosive and toxic.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR