Hydrion Test Paper Strips
Supplier: Micro Essential
165A 165B 165A 165B
60775-203CS 52.05 CAD
60775-203 60775-225 470200-342 470200-344
Hydrion Test Paper Strips
Test Papers pH Indicator Papers and Test Strips
Use Hydrion A to test in the range of 2-4-6-8-10 and Hydrion B for the range of 1-3-5-7-9-11. Both strips let you test the pH scale in 2.0 units.
Includes: color chart.
Hydrion 1-12 pH Paper is a general use pH paper that can be used for: 
Classroom demonstrations, and general laboratories.
Order Now
SPECIFICATIONS
Environmentally Preferable