Ward's® Horizontal Gel Box System

Supplier: LABNET INTERNATIONAL CA
E1015-10-VWR E1010-10-WAR E1007-10-WAR
470020-218EA 794.56 CAD
470020-218 470144-930 470145-450
Ward's® Horizontal Gel Box System
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Designed for Ease of Use, Safety and Durability.

These gel tanks are molded for leak-proof performance and a long service life. To cast, simply snap the rubber gates onto the ends of the UV transparent gel tray, place the comb in position and pour the gel- no taping is necessary. Slots on the sides of the gel trays hold the combs in place. The two-piece, 1.5 mm thick combs can be adjusted to the depth of the well within the gel. After the gel has hardened, the combs and gates are removed and the gel and tray placed in the gel box. Loading strips may be placed on the bottom of the gel box to aid in well visualization. The corrosion resistant platinum electrodes are located in cassettes to protect them during cleaning and provide for easy replacement. A drip ring on the lid recycles condensed buffer and prevents it from dripping directly onto the gel. Power leads are attached to the gel box lid, ensuring that power is connected only when the lid is properly in place; power is cut off when lid is removed. Side tabs and pressure pads make lid removal easy and prevent sloshing of buffer.

Specifications:
Buffer Volume 225mL
Maximum Sample Capacity: 64
Gel Size 7 x 10cm/10 x 10cm
Unit Dimensions 9 x 21 x 9cm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR