Hooks Collars
Supplier: GSC International
HKCLR-13 HKCLR-13-C HKCLR-19-C HKCLR-19
470016-118EA 10.75 CAD
470016-118 470352-508 470352-506 470352-980
Hooks Collars
Clamps General Purpose Clamps
The Hooks Collar is constructed with a ring that fits over a support stand rod, a hook and thumb screw.
  • Metal ring
  • Metal hook
  • Different sizes and materials to fit all classroom needs and budgets
The Hooks Collar is used in a physics classroom for suspending items such as pulleys, cords, or weights from a rod or crass bar.
Caution: Prop 65 Lead Warnings are attached to the package.