High-Low Alcohol Burner Stand
Supplier: Shiv Dial Sud & Sons
CH10654 CH10654
470149-522CS 2145.5 CAD
470149-522 470200-546
High-Low Alcohol Burner Stand
Tripods
Perfect for any style alcohol lamp, this stand can be used in two different height levels (9 or 13 cm) by simply removing the wire gauze top, turning the stand on its side, and putting the top back on. It is constructed of stainless steel rod with an 8 x 10 cm wire gauze surface. The edges of the wire gauze are turned under for safety.