High Form Porcelain Crucibles
Supplier: SILICO & CHEMICO PORCELAIN SE
J241 J240 J238 J237 J232 J239
470148-910CS 298.4 CAD
470148-910 470148-954 470149-562 470149-564 470148-980 470149-078
High Form Porcelain Crucibles
Crucibles
These crucibles are similar to wide-form porcelain crucibles but taller.
  • Without covers
  • Max. use temp. is 1050 Celsius
  • Rate of cooling and heating should not increase 200 degrees per hour
They don't have covers.
Order Now
SPECIFICATIONS
Environmentally Preferable