Hexanes

Supplier: ALDON CORP SE

HH0025-500ML HH0025-100MLR HH0025-500MLR HH0025-PK/2R
470301-206EA 60.05 CAD
470301-206 470301-208 470301-210 470301-212
Hexanes
Chemicals for Science Education

CAS Number: 110-54-3
Formula: C6H14
Density: 0.660—0.690 g/mL
Boiling Point: 63—68°C
Solubility: Alcohol, Acetone, and Ether
Synonyms: Hexane and n-Hexane
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is flammable and toxic.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR