EDVOTEK Hexagel™ M36 Electrophoresis Chamber
Supplier: Edvotek
515 680-A 684-A
470005-306EA 772.6 CAD
470005-306 470005-310 470005-314 470005-312
EDVOTEK Hexagel™ M36 Electrophoresis Chamber
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Run six agarose gels simultaneously.
  • Lifetime warranty
  • Contoured lid for enhanced gel visualization
  • Large color coded push tabs for easy lid insertion and removal
  • Pour spout for buffer disposal
  • User replaceable electrodes
Apparatus supports up to six groups of students. Produces excellent results in 30 to 40 minutes.
The chamber itself is seamless and made with injection-molded UV transparent construction and adjustable leveling feet. It comes with six combs that allow you to make six wells per gel and requires 1000 mL of buffer.