Herbarium Paste
Supplier: Avantor
WARD470021-054
470021-054EA 47.3 CAD
470021-054
Herbarium Paste
Adhesives
Specially formulated to adhere plant specimens to mounting sheets.
  • Waterbased
  • Easy cleanup
  • Low odor
This longtime industry standard paste is waterbased for easy cleanup and low odor. The dispenser tip measures just the right amount of paste to attach herbarium specimens to mounting sheets. One pint bottle.