Guar Gum

Supplier: ALDON CORP SE

GG0246-10G_U GG0246-100G GG0246-500G
470301-194EA 14 CAD
470301-194 470301-196 470301-198
Guar Gum
Chemicals for Science Education

CAS Number: 9000-30-0
Solubility: Water
Synonyms: Guar Flour
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR