Graphite

Supplier: ALDON CORP SE

ADGG226250 GG0225-500G
470109-914EA 101.05 CAD
470109-914 470301-190
Graphite
Chemicals for Science Education

CAS Number: 7782-42-5
Formula Weight: 12.01
Formula: C
Density (g/mL): 2.26
Freezing Point (°C): Sublimes
Synonyms: Carbon, Black Lead
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR