Gram's Iodine

Supplier: ALDON CORP SE

IX0180-1L IX0180-500ML GG0211-100ML IX0180-100ML
470301-184EA 146.3 CAD
470301-184 470301-188 470301-182 470301-186
Gram's Iodine
Chemicals for Science Education

CAS Number: Mixture
Density: 1.0 g/mL
Boiling Point: 100 °C
Freezing Point: 0 °C
Synonyms: Iodine-Potassium Iodide Solution, IKI, Iodine-Iodine, Iodine Solution, Iodine Lugol's Dilute, and Iodine 2%
Shelf Life: 36 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR