Polypropylene Cylinders with Handle
Supplier: Bel-Art
F28461-2000_SE F28461-1000_SE
470003-612EA 166.85 CAD
470003-612 470012-892
Polypropylene Cylinders with Handle
Cylinders Measuring Cylinders
Put an End to the Difficult Task of Pouring from Large, Heavy Cylinders.
  • Double scale raised graduations
  • Chemically resistant
  • Autoclavable to 121°C