Gloeotrichia
Supplier: VWR
L127
470179-882EA 48.9 CAD
470179-882 470179-880
Gloeotrichia
Specimens Live Specimens Bacteria - Living Material
Freshwater. Filamentous. Trichomes radiating from basil heterocysts. Basic culture solution.
Shipped in a form that provides the best quality specimen. Most are supplied in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars.