Glo Germ
Supplier: Glo Germ
OIL POWDER BRHP GEL
470004-260EA 38.05 CAD
470004-260 470004-262 470100-682 470100-620
Glo Germ
Educational Materials Biology Educational Materials Physiology Learning Activities
Used Worldwide to Teach Proper Handwashing Technique.
  • Choose Powder, Liquid, or Gel
  • Test Handwashing or Cleaning Effectivity
  • Invisible Until Exposed to UV Light
Both the oil and the powder contain minute pieces of plastic that glow under UV light. To become germ detectives, students apply a small amount of oil before handwashing and then try to spot remaining “germs” under a UV light. To investigate the thoroughness of surface cleaning, students wipe away a light dusting of Glo Germ powder and hunt for germs with the UV light.
Glo Germ gel isMore Product Information