Glass-Reinforced Acetyl Test Tube Racks
Supplier: Ward's Science
202112-W 202113-W 202110-W 202111-W
470148-866CS 328.25 CAD
470148-866 470149-102 470149-126 470149-130
Glass-Reinforced Acetyl Test Tube Racks
Racks Tube Racks
Perfect for Submersion.
  • Heavy-duty rack does not float
  • Withstands temperatures to 150 °C
  • Autoclavable and chemical resistant