Glass Vials with Screw Caps
Supplier: Glass Vials
V1760C-FM V2385C-FM V1235C-FM V2170C-FM
470206-380CS 229.85 CAD
470206-380 470206-384 470206-388 470206-390
Glass Vials with Screw Caps
Vials
Ideal for a variety of uses.
  • Accompanied by a polypropylene foam lined closed top cap
  • Cap designed with a continuous thread 'CT' finish
  • Creates a positive seal
  • Available in a wide range of sizes