Glass Tubing Manipulator
Supplier: JET RUBBER COMPANY
6199000
470016-798PR 212.75 CAD
470016-798
Glass Tubing Manipulator
Puller and Inserter Tools
Protects hands by eliminating accidents when inserting glass tubing into cork or rubber stoppers.
  • Made from hard rubber with large shield in front
  • Overall diameter is 76 mm