SP Wilmad-LabGlass Glass Separatory Funnels, SP Industries
Supplier: SP Industries
LG-8420-106 LG-8420-108
470122-372EA 208.6 CAD
470122-372 470122-374
SP Wilmad-LabGlass Glass Separatory Funnels, SP Industries
Funnels Separating Funnels
  • T-Solid Stopper
  • Borosilicate Glass
Ordering information: Supplied complete with glass stopcock plug and glass pennyhead flask length stopper.