Glass Depression Slides
Supplier: United Scientific Supplies
CSTK01 CS3X12-PK/12 4-13057-DZ-12
470019-022PK 19.45 CAD
470019-022 470200-930 470235-728
Glass Depression Slides
Slides Microscope Slides
These depression slides are ideal for viewing living specimens suspended in liquid.
  • Made with quality sheet glass with ground edges
  • Sold in boxes of 12 slides
  • Available with single or double concavities
These precleaned slides have a round, shallow depression in the center of the slide, making it easy to view materials in liquid.
Cavities measure approximately 16mm in diameter, and 0.8mm deep.
Order Now
SPECIFICATIONS
Environmentally Preferable