Ward's® Giant Toad (Bufo marinus)
Supplier: BRUCE EDELMAN
878260 0
470176-522EA 102.6 CAD
470176-522 470189-368
Ward's® Giant Toad (Bufo marinus)
Specimens Live Specimens Animals - Living Material
This toad thrives with minimum attention and may be housed in a terrarium with access to water and dry land.
  • Low maintenance
  • Excellent for physiology studies
  • Exhibits territoriality and defensive behavior
Feed live insects such as crickets.
Size ranges from 10 to 13 cm.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.