Geoblox Topographic Landform Models Sets
Supplier: JOHN HAROLD KOONZ
47321-04 47321-03
470004-722EA 55.85 CAD
470004-722 470004-724
Geoblox Topographic Landform Models Sets
Models Geology Models
Geoblox models provide a multi-learning approach to help students understand concepts and structures from the inside out as they build their own models. Cardboard and building supplies are not included.
  • Thirty Models in set one
  • Twenty Models in set two
  • Multi-Learning Approach
  • Comb-Bound Set
  • Multiple Uses for Each Set
  • Descriptions, Sketch, Instructions, and Questions Included
Geoblox models provide a multi-learning approach to helping students understand a concept or a structure from the inside out as they build their own models. Activities, such as drawing diagrams of models before they’re built, provide opportunities for students to develop thinking skills.
NOTE: Models do not come preassembled. Students and educators are meant to use the patterns and instructions in the booklet to construct the models for educational purposes. Constructed models can then be used as a permanent classroom display.
Each set of model patterns is comb bound and can be used as a copymaster, which produces enough material for an entire class. Models can also be enlarged for use as demonstration models.
Ordering information: For Set 1, the 30 topographic maps of landforms each feature a description, sketch, instructions, and two questions. Models include an atoll, emergent highland coast, estuary, sea stacks, spit, submarine canyon, tombolo, drumlins, eskers, iceberg, alluvial fan, badlands, butte, playa lake, cinder cone, entrenched meander, exfoliation dome, flood basalt, volcanic plug, earth dam, impact crater, karst, stair step topography, alpine glacier, cirque lake, horn, dendritic drainage, entrenched meanders, levees, meanders scars, and waterfall.
For Set 2, the 20 topographic maps of landforms each feature a description, sketch, instructions, and two questions. Cardboard and building supplies are not included. Models include a Fjord, Ria Coastline, Truncated Headland, Bajada, Cut and Fill (Before), Cut and Fill (After), Anticlinal Mountain, Continental Divide, Escarpment, Water Gap, Wind Gap, Braided Stream, Delta, Radial Drainage, Springs, Stream Piracy, Terraced Rivers, Trellis Drainage