Gelatin

Supplier: ALDON CORP SE

GG0025-500G GG0025-500GP
470301-132EA 93.7 CAD
470301-132 470301-134
Gelatin
Chemicals for Science Education

CAS Number: 9000-70-8
Formula Weight: Mixture
Formula: Natural Product
Density (g/mL): >1.0
Boiling Point (°C): Decomposes
Freezing Point (°C): Decomposes
Solubility: Hot Water
Shelf Life (months): 36
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR