Geissler Burets with Teflon® Stopcock Plugs
Supplier: Thermo Fisher Scientific
3650-0050 3650-0050 3650-0025
470200-450CS 1270.3 CAD
470200-450 470203-504 470211-804
Geissler Burets with Teflon® Stopcock Plugs
Burettes
Geissler burets have easy-to-read graduations and sturdy stopcocks.
  • Clear Acrylic
  • Economy-Grade
  • Leakproof
  • Easy-to-Read, Blue-Enameled Graduations
  • Self-Lubricating Teflon
The stopcock and tip are made of durable polymethylpentene (PMP), and the stopcock plug is made of Leakproof, self-lubricating Teflon.
The burets are unaffected by most dilute acids and bases but are not recommended for use with alcohol or organic solvents.