Funnel Support
Supplier: VWR International
CH0689D
470123-524EA 62.25 CAD
470123-524
Funnel Support
Retort Stands
With iron clamp for support rods of any size to 5/8". The four holes are beveled and slotted and are spaced at 41/2" center distance.