Funnel Sets, Rochester
Supplier: Avantor
248
470014-960KT 64.8 CAD
470014-960 470175-294
Funnel Sets, Rochester
Funnels Filtration Funnels Filter Funnels
Each set includes three funnels of different sizes. Small funnels are made of polypropylene; large funnels are made of polyethylene. Sizes in small funnel set: 60 mm top dia. x 20 mm stem, 80 mm top dia. x 22 mm stem, 110 mm top dia. x 36 mm stem. Sizes in large set: 127 mm top dia. x 45 mm stem, 160 mm top dia. x 50 mm stem, 200 mm top dia. x 60 mm stem.