Freeze-Dried Grassfrogs
Supplier: Avantor
470000-958PK 1132.8 CAD
470000-958 470001-394 470001-396 470001-398 470001-400
Freeze-Dried Grassfrogs
Specimens Preserved Specimens Preserved Animal Materials
Freeze-dried grassfrogs. Available in vacuum packs.
  • Amphibian anatomy
  • Economical
Our freeze-dried specimens, which use no chemicals, can be rehydrated in 10% alcohol for dissection.
Order Now
SPECIFICATIONS
Environmentally Preferable