Forward Vision™ Four Student Workstations
Supplier: DIVERSIFIED WOODCRAFTS INC SE
2916K 2936K 2956KF
470107-562EA 18491.65 CAD
470107-562 470107-558 470107-564
Forward Vision™ Four Student Workstations
Furniture Tables and Workbenches
All in one: classroom table and lab bench. The ForwardVision™ workstation allows up to four students to work at a single station while all are facing the teacher.
  • 1" solid epoxy resin surface
  • Constructed of solid oak and oak veneers
  • Rubber base molding and durable chemical resistant UV finish
  • All doors and drawers with lock
  • Two models available: door cabinet with two adjustable shelves or the door/drawer cabinet with one adjustable shelf and a drawer
This popular workstation meets the needs of students and teachers today. The unique design leaves enough room to store four stools (not included) under the workstation.
Sink dimensions: 22³/₈W×15L×17" D
Certifications: SEFA compliant and MAS Certified Green.
Delivery information: Unit comes with a sink, two combination water/gas fixtures, two GFI electrical outlets and AC duplex receptacles, and ample storage behind two locking door/drawer cabinets with one adjustable shelf and a drawer. The 2916KLH and 2936KLH include two rod sockets and LabHands for holding laptop computers and books.