Formaldehyde

Supplier: ALDON CORP SE

FF0250-3.8L FF0250-3.8LR FF0250-500MLR FF0269-3.8L FF0268-500MLR
470301-106EA 68.25 CAD
470301-106 470301-108 470301-110 470301-112 470301-116
Formaldehyde
Chemicals for Science Education

CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Toxic, Irritant, Suspected Human Carcinogen
Density (g/mL): 1.080
Boiling Point (°C): 96
Solubility: Water and Alcohol
Synonyms: Formalin Solution
Shelf Life (months): 12
Storage: White


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR